Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexylacetic acid 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11127-0050 11127-0250 11127-1000
TS11127-0050EA 43 USD InStock
TS11127-0050 TS11127-0250 TS11127-1000
Cyclohexylacetic acid 98+%
Cyclohexylacetic acid


Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 242
Melting Pt: 28…32 °C
Density: 1.007 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001518
CAS Number: 5292-21-7
EINECS: 226-132-0
Order Now
Specification Test Results
Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 27 to 31 °C
Titration with NaOH ≥98.0 %
Identification Passes test
Sensitivity (to amino acids) Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty