Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tricarballylic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

13936-0050 13936-0500
TS13936-0050EA 45 USD InStock
TS13936-0050 TS13936-0500
Tricarballylic acid 99%
Tricarballylic acid


Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 266
Melting Pt: 160
Storage Temperature: Ambient
MDL Number: MFCD00002723
CAS Number: 99-14-9
EINECS: 202-733-3
Merck Index: 12,09752
Order Now
Specification Test Results
Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 154 to 159 °C
Titration with TBAH ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty