Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,6-Heptadien-4-ol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15009-0050 15009-0250
TS15009-0050EA 104 USD InStock
TS15009-0050 TS15009-0250
1,6-Heptadien-4-ol 97%
1,6-Heptadien-4-ol


Formula: C₇H₁₂O
Boiling Pt: 151 °C
Density: 0.86 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00008663
CAS Number: 2883-45-6
EINECS: 220-742-0
UN: 1987
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4530 (20°C, 589 nm)
GC ≥96.0 %
Residue after ignition 35 % to 50 %
pH 5 to 8 (1 % solution)
Specific optical rotation +75° to +105° (20°C, 589 nm) (6% in water)
Specific viscosity 0.018 to 0.032 (1.0M in NaCl, 25°C)
Sulfur (S) 16 to 20 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty