Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl cyanoacetate 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11828-0010 11828-0051 11828-2500
TS11828-0010EA 101 USD InStock
TS11828-0010 TS11828-0051 TS11828-2500
Ethyl cyanoacetate 98+%
Ethyl cyanoacetate


Formula: C₅H₇NO₂
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1.06 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001940
CAS Number: 105-56-6
EINECS: 203-309-0
Merck Index: 13,03821
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.0 %
Color scale ≤50 APHA
Free acid ≤10 mg/kg (as cyanoacetic acid)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty