Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl cyanoacetate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

14864-0010 14864-2500
TS14864-0010EA 99 USD InStock
TS14864-0010 TS14864-2500
Methyl cyanoacetate 97%
Methyl cyanoacetate


Formula: C₄H₅NO₂
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.12 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00001939
CAS Number: 105-34-0
EINECS: 203-288-8
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20°C, 589 nm)
GC ≥96.0 %
Water ≤5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty