Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl chloroacetate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

33616-0050 33616-0250
TS33616-0050EA 29 USD InStock
TS33616-0050 TS33616-0250
Benzyl chloroacetate 98%
Benzyl chloroacetate


Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 132 °C (12 mmHg)
Density: 1.22 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00157062
CAS Number: 140-18-1
EINECS: 205-400-0
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5240 to 1.5270 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty