Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxybenzamide 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12572-0050 12572-0250
TS12572-0050EA 35 USD InStock
TS12572-0050 TS12572-0250
4-Methoxybenzamide 97%
4-Methoxybenzamide


Formula: C₈H₉NO₂
Boiling Pt: 295 °C (760 mmHg)
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00007995
CAS Number: 3424-93-9
EINECS: 222-319-6
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 164 to 167 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty