Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Diphenylphosphino)benzoic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

38293-0010 38293-0050
TS38293-0010EA 78 USD InStock
TS38293-0010 TS38293-0050
2-(Diphenylphosphino)benzoic acid 98%
2-(Diphenylphosphino)benzoic acid


Formula: C₁₉H₁₅O₂P
Melting Pt: 177…181 °C
MDL Number: MFCD00674024
CAS Number: 17261-28-8
EINECS: 241-293-7
Order Now
Specification Test Results
Appearance Light yellow Powder
Infrared spectrum Conforms
Melting point 177 to 181 °C
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty