Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Piperidine-N-carbaldehyde 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11973-0010 11973-2500
TS11973-0010EA 232 USD InStock
TS11973-0010 TS11973-2500
Piperidine-N-carbaldehyde 99%
Piperidine-N-carbaldehyde


Formula: C₆H₁₁NO
MW: 113.16 g/mol
Melting Pt: –31 °C
Density: 1.019 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00006483
CAS Number: 2591-86-8
EINECS: 219-986-0
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4850 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty