Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Furohydrazide 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15716-0050 15716-0250
TS15716-0050EA 40 USD InStock
TS15716-0050 TS15716-0250
2-Furohydrazide 98%
2-Furohydrazide


Formula: C₅H₆N₂O₂
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00003235
CAS Number: 3326-71-4
EINECS: 222-046-2
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Beige Crystals
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty