Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl acetylenedicarboxylate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

29635-0250 29635-1000
TS29635-0250EA 82 USD InStock
TS29635-0250 TS29635-1000
Diethyl acetylenedicarboxylate 97%
Diethyl acetylenedicarboxylate


Formula: C₈H₁₀O₄
MW: 170.16 g/mol
Boiling Pt: 107…110 °C (11 mmHg)
Melting Pt: 1…3 °C
Flash Pt: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009186
CAS Number: 762-21-0
EINECS: 212-095-8
UN: 1760
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4430 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty