Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl mercaptoacetate 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

14888-1000 14888-5000
TS14888-1000EA 33 USD InStock
TS14888-1000 TS14888-5000
Methyl mercaptoacetate 95%
Methyl mercaptoacetate


Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 156…159 °C (10 mmHg)
Melting Pt: –24 °C
Density: 1.187 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00004873
CAS Number: 2365-48-2
EINECS: 219-121-7
UN: 1228
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4640 to 1.4670 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty