Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triethyl methanetricarboxylate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

13963-0250 13963-1000
TS13963-0250EA 130 USD InStock
TS13963-0250 TS13963-1000
Triethyl methanetricarboxylate 98%
Triethyl methanetricarboxylate


Formula: C₁₀H₁₆O₆
Boiling Pt: 253 °C (760 mmHg)
Melting Pt: 29 °C
Density: 1.1 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009150
CAS Number: 6279-86-3
EINECS: 228-477-2
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4260 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty