Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium propionate 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

40390-0010 40390-0050 40390-0251
TS40390-0010EA 62 USD InStock
TS40390-0010 TS40390-0050 TS40390-0251
Calcium propionate 98+%
Calcium propionate


Formula: (CH₃CH₂COO)₂Ca
MW: 186.22 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00036137
CAS Number: 4075-81-4
EINECS: 223-795-8
Merck Index: 13,01703
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration Complexometric ≥98.0 % (on dry substance)
Loss on drying ≤4 % (105°C, 2 Hrs)
pH 6.9 to 9.0 (10 % solution)
Fluoride (F) ≤10 ppm
Arsenic (As) ≤3 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.3 % (in water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty