Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl cinnamate 98% (trans-isomers)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

15053-0025 15053-1000 15053-5000
TS15053-0025EA 371 USD InStock
TS15053-0025 TS15053-1000 TS15053-5000
Ethyl cinnamate 98% (trans-isomers)
Ethyl cinnamate


Formula: C₁₁H₁₂O₂
Boiling Pt: 271 °C (760 mmHg)
Melting Pt: 6.5…7.5 °C
Density: 1.046 g/cm³
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00009189
CAS Number: 103-36-6
EINECS: 203-104-6
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5580 to 1.5610 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25/25°C) 1.045 to 1.048
Acid value ≤1 mg KOH/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty