Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium methyl sulphate 99% pract.

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

41490-0250 41490-1000
TS41490-0250EA 123 USD InStock
TS41490-0250 TS41490-1000
Sodium methyl sulphate 99% pract.
Sodium methyl sulphate


Formula: CH₃NaO₄S
MW: 134.09 g/mol
Melting Pt: 193
Storage Temperature: Ambient
MDL Number: MFCD00013457
CAS Number: 512-42-5
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty