Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium octadecyl sulphate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

38489-0010 38489-0050 38489-0250
TS38489-0010EA 78 USD InStock
TS38489-0010 TS38489-0050 TS38489-0250
Sodium octadecyl sulphate 98%
Sodium octadecyl sulphate


Formula: C₁₈H₃₇NaO₄S
Melting Pt: 211…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00007469
CAS Number: 1120-04-3
Order Now
Specification Test Results
Appearance White to light yellow to light beige Powder
Infrared spectrum Conforms
Loss on drying ≤7 %
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty