Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4-Dimethyl-1-pentene 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

42627-0010 42627-0050 42627-0250
TS42627-0010EA 39 USD InStock
TS42627-0010 TS42627-0050 TS42627-0250
4,4-Dimethyl-1-pentene 99%
4,4-Dimethyl-1-pentene


Formula: C₇H₁₄
MW: 98.19 g/mol
Boiling Pt: 73
Melting Pt: -136.6
Density: 0.682 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00026337
CAS Number: 762-62-9
UN: 3295
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3910 to 1.3930 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty