Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,4,5-Tetramethylbenzene 97+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

13837-0050 13837-1000 13837-5000
TS13837-0050EA 25 USD InStock
TS13837-0050 TS13837-1000 TS13837-5000
1,2,4,5-Tetramethylbenzene 97+%
1,2,4,5-Tetramethylbenzene


Formula: C₁₀H₁₄
Boiling Pt: 196…197 °C (760 mmHg)
Melting Pt: 78…82 °C
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00008528
CAS Number: 95-93-2
UN: 1325
ADR: 4.1,II
Order Now
Specification Test Results
Appearance Semitransparent white to brown Crystals
Infrared spectrum Conforms
Melting point 78 to 82 °C
GC ≥97.0 %
Elemental Analysis 4.5 to 4.9 % N

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty