Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrophenylboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43389-0010 43389-0050
TS43389-0010EA 68 USD InStock
TS43389-0010 TS43389-0050
4-Nitrophenylboronic acid 97%
4-Nitrophenylboronic acid


Formula: C₆H₆BNO₄
MW: 166.93
Melting Pt: 270…271 °C
Storage Temperature: Ambient
MDL Number: MFCD00161360
CAS Number: 24067-17-2
Order Now
Specification Test Results
Appearance White to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty