Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl myristate 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

38554-0250 38554-1000 38554-5000
TS38554-0250EA 29 USD InStock
TS38554-0250 TS38554-1000 TS38554-5000
Ethyl myristate 98+%
Ethyl myristate


Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 295 °C (760 mmHg)
Melting Pt: 11…12 °C
Density: 0.861 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008984
CAS Number: 124-06-1
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty