Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Bromoacetanilide 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43633-0050 43633-0250
TS43633-0050EA 56 USD InStock
TS43633-0050 TS43633-0250
2'-Bromoacetanilide 96%
2'-Bromoacetanilide


Formula: H₃CC(O)NH(C₆H₄Br)
Melting Pt: 100…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00099252
CAS Number: 614-76-6
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 103 °C
GC ≥95.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty