Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl bromoacetate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15434-0100 15434-0500 15434-2500
TS15434-0100EA 53 USD InStock
TS15434-0100 TS15434-0500 TS15434-2500
tert-Butyl bromoacetate 99%
tert-Butyl bromoacetate


Formula: BrCH₂COOC(CH₃)₂
MW: 195.06 g/mol
Boiling Pt: 50
Density: 1.338 g/cm³
Flash Pt: 49 °C
MDL Number: MFCD00000188
CAS Number: 5292-43-3
EINECS: 226-133-6
UN: 1993
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty