Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl bromoacetate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15859-0025 15859-1000 15859-5000
TS15859-0025EA 429 USD InStock
TS15859-0025 TS15859-1000 TS15859-5000
Ethyl bromoacetate 98%
Ethyl bromoacetate


Formula: BrCH₂COOCH₂CH₃
MW: 167 g/mol
Boiling Pt: 159
Melting Pt: -38
Density: 1.506 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00000191
CAS Number: 105-36-2
EINECS: 203-290-9
UN: 1603
ADR: 6.1 II
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4500 to 1.4520 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤300 APHA
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.002 %
Insoluble matter ≤0.01 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty