Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propyl propionate 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

38552-0010 38552-0500 38552-2500
TS38552-0010EA 87 USD InStock
TS38552-0010 TS38552-0500 TS38552-2500
Propyl propionate 98+%
Propyl propionate


Formula: C₆H₁₂O₂
Boiling Pt: 123 °C (760 mmHg)
Melting Pt: –76 °C
Density: 0.881 g/cm³
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009373
CAS Number: 106-36-5
UN: 3272
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3820 to 1.3940 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty