Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl acrylate 99% stabilised

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

12619-0010 12619-0025 12619-0100 12619-0101 12619-5000
TS12619-0010EA 65 USD InStock
TS12619-0010 TS12619-0025 TS12619-0100 TS12619-0101 TS12619-5000
Methyl acrylate 99% stabilised
Methyl acrylate (monomer)


Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.956 g/cm³
Flash Pt: –3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008627
CAS Number: 96-33-3
EINECS: 202-500-6
UN: 1919
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4010 to 1.4040 (20°C, 589 nm)
Water ≤0.05 %
GC ≥98.5 %
Color scale ≤10 APHA
Stabilizer 10 to 20 ppm MEHQ
Acidity ≤0.0004 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Calcium (Ca) ≤0.5 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Peroxides ≤0.005 % (as H2O2)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty