Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Butylboronic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17238-0010 17238-0050 17238-0250
TS17238-0010EA 33 USD InStock
TS17238-0010 TS17238-0050 TS17238-0250
1-Butylboronic acid 98%
1-Butylboronic acid


Formula: C₄H₁₁BO₂
Melting Pt: 94…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002106
CAS Number: 4426-47-5
EINECS: 224-607-7
Order Now
Specification Test Results
Appearance White to off-white Glistening powder or flakes
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty