Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

31222-0010 31222-0050 31222-2500
TS31222-0010EA 184 USD InStock
TS31222-0010 TS31222-0050 TS31222-2500
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid


Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
Boiling Pt: 116…118 °C (1.5 mmHg)
Melting Pt: 46…49 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004184
CAS Number: 20445-31-2
EINECS: 243-829-5
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Viscous liquid or low melting solid
Infrared spectrum Conforms
Specific optical rotation +71° to +74° (20°C, 589 nm) (c=2, CH3OH)
Titration with NaOH 98.5 to 101.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty