Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminobenzenesulphonic acid 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

40064-0250 40064-1000 40064-5000
TS40064-0250EA 23 USD InStock
TS40064-0250 TS40064-1000 TS40064-5000
2-Aminobenzenesulphonic acid 95%
2-Aminobenzenesulphonic acid


Formula: C₆H₇NO₃S
MW: 173.19 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007705
CAS Number: 88-21-1
EINECS: 201-810-9
UN: 2585
ADR: 8 III
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥94 %
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty