Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Bis(hydroxymethyl)propionic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

14797-0010 14797-0050 14797-5000
TS14797-0010EA 77 USD InStock
TS14797-0010 TS14797-0050 TS14797-5000
2,2-Bis(hydroxymethyl)propionic acid 98%
2,2-Bis(hydroxymethyl)propionic acid


Formula: C₅H₁₀O₄
Melting Pt: 185…190 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004199
CAS Number: 4767-03-7
EINECS: 225-306-3
Order Now
Specification Test Results
Appearance White Powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty