Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester 97+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

38296-0010 38296-0050 38296-0250
TS38296-0010EA 73 USD InStock
TS38296-0010 TS38296-0050 TS38296-0250
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester 97+%
1-Methyl-1H-pyrazole-4-boronic acid pinacol ester


Formula: C₁₀H₁₇BN₂O₂
Melting Pt: 62…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03789259
CAS Number: 761446-44-0
Order Now
Specification Test Results
Appearance Off-white to yellow Crystals
Infrared spectrum Conforms
Melting point 62 to 65 °C
GC ≥97.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty