Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Diethynylbenzene 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39155-0010 39155-0050
TS39155-0010EA 71 USD InStock
TS39155-0010 TS39155-0050
1,4-Diethynylbenzene 95%
1,4-Diethynylbenzene


Formula: C₁₀H₆
Melting Pt: 94…98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078375
CAS Number: 935-14-8
Order Now
Specification Test Results
Appearance White to orange to brown Crystalline powder, crystals and/or chunks
Infrared spectrum Conforms
Melting point 90 to 96 °C
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty