Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethylene glycol dimethacrylate (ethylene dimethacrylate) 98% stabilised

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

40992-0010 40992-0051 40992-2500
TS40992-0010EA 163 USD InStock
TS40992-0010 TS40992-0051 TS40992-2500
Ethylene glycol dimethacrylate (ethylene dimethacrylate) 98% stabilised
Ethylene glycol dimethacrylate (ethylene dimethacrylate)


Formula: C₁₀H₁₄O₄
MW: 198.21 g/mol
Boiling Pt: 98…100 °C (5 mmHg)
Melting Pt: –40 °C
Density: 1.05 g/cm³
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008590
CAS Number: 97-90-5
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4530 to 1.4550 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 90 to 150 ppm MEHQ

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty