Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methoxycarbonyl)phenylboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36983-0010 36983-0050 36983-0250
TS36983-0010EA 48 USD InStock
TS36983-0010 TS36983-0050 TS36983-0250
3-(Methoxycarbonyl)phenylboronic acid 97%
3-(Methoxycarbonyl)phenylboronic acid


Formula: C₈H₉BO₄
Melting Pt: 187…191 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093046
CAS Number: 99769-19-4
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty