Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Quinolineboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43020-0010 43020-0050
TS43020-0010EA 145 USD InStock
TS43020-0010 TS43020-0050
6-Quinolineboronic acid 97%
6-Quinolineboronic acid


Formula: C₉H₈BNO₂
Melting Pt: 144…145 °C
Storage Temperature: Ambient
MDL Number: MFCD03093100
CAS Number: 376581-24-7
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥99.0 %
Color scale ≤25 APHA
Stabilizer Approx. 50 ppm BHT
Non-volatile matter ≤0.005 %
Acidity ≤0.0004 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Calcium (Ca) ≤0.5 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Peroxides ≤0.005 % (as H2O2)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty