Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Aminomethyl)phosphonic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

29164-0010 29164-0050 29164-2500
TS29164-0010EA 161 USD InStock
TS29164-0010 TS29164-0050 TS29164-2500
(Aminomethyl)phosphonic acid 99%
(Aminomethyl)phosphonic acid


Formula: CH₆NO₃P
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008105
CAS Number: 1066-51-9
UN: 3261
ADR: 8,III
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty