Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl benzylphosphonate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11417-0250 11417-1000 11417-5000
TS11417-0250EA 80 USD InStock
TS11417-0250 TS11417-1000 TS11417-5000
Diethyl benzylphosphonate 99%
Diethyl benzylphosphonate


Formula: C₁₁H₁₇O₃P
MW: 228.23 g/mol
Boiling Pt: 106…108 °C (1 mmHg)
Density: 1.09 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009078
CAS Number: 1080-32-6
EINECS: 214-097-4
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.4980 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty