Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzylphosphonic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

39334-0010 39334-0050 39334-0250
TS39334-0010EA 36 USD InStock
TS39334-0010 TS39334-0050 TS39334-0250
Benzylphosphonic acid 98%
Benzylphosphonic acid


Formula: C₇H₉O₃P
MW: 172.11 g/mol
Melting Pt: 165…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00039519
CAS Number: 6881-57-8
UN: 3261
ADR: 8 III
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Melting point 169 to 173 °C
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty