Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetohydroxamic acid 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17013-0050 17013-0250
TS17013-0050EA 57 USD InStock
TS17013-0050 TS17013-0250
Acetohydroxamic acid 98+%
Acetohydroxamic acid


Formula: C₂H₅NO₂
MW: 75.0666 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00009994
CAS Number: 546-88-3
EINECS: 208-913-8
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 85 to 90 °C
GC ≥98.0 %
Titration with NaOH 98.0 to 102.0 %
Specific optical rotation -24° to -10° (20 °C, 589 nm) (c=1,H2O)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty