Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-bromocrotonate 75% tech.

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11812-0050 11812-0250 11812-1000
TS11812-0050EA 31 USD InStock
TS11812-0050 TS11812-0250 TS11812-1000
Ethyl-4-bromocrotonate 75% tech.
Ethyl-4-bromocrotonate


Formula: C₆H₉BrO₂
MW: 193.05 g/mol
Boiling Pt: 94…95 °C (12 mmHg)
Density: 1.4 g/cm³
Flash Pt: 97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000247
CAS Number: 37746-78-4
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
GC ≥70 %
Thiocyanate (SCN) ≤200 ppm
Phosphate (PO4) ≤200 ppm
Sulfate (SO4) ≤500 ppm
Lead (Pb) ≤5 ppm
Sulfur (S) ≤50 ppm
Iron, total (as Fe) ≤0.005 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty