Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Pentynyl)phthalimide 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42573-0050 42573-0250
TS42573-0050EA 67 USD InStock
TS42573-0050 TS42573-0250
N-(4-Pentynyl)phthalimide 97%
N-(4-Pentynyl)phthalimide


Formula: C₁₃H₁₁NO₂
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD06798110
CAS Number: 6097-07-0
Order Now
Specification Test Results
Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 87 to 91 °C
GC ≥96.0 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty