Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenylcyclopentanecarboxylic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16114-0050 16114-0250
TS16114-0050EA 26 USD InStock
TS16114-0050 TS16114-0250
1-Phenylcyclopentanecarboxylic acid 98%
1-Phenylcyclopentanecarboxylic acid


Formula: C₁₂H₁₄O₂
Melting Pt: 157…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00001372
CAS Number: 77-55-4
EINECS: 201-037-7
Order Now
Specification Test Results
Appearance Cream to grey-brown Powder
Infrared spectrum Conforms
Melting point 157 to 161 °C
GC ≥97.5 %
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty