Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenylcyclopropanecarboxylic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

17068-0100 17068-0500
TS17068-0100EA 79 USD InStock
TS17068-0100 TS17068-0500
1-Phenylcyclopropanecarboxylic acid 97%
1-Phenylcyclopropanecarboxylic acid


Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00001288
CAS Number: 6120-95-2
EINECS: 228-090-9
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 84 to 87 °C
GC ≥96.0 %
Water ≤0.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty