Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl dimethoxyacetate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

41418-0050 41418-0250
TS41418-0050EA 45 USD InStock
TS41418-0050 TS41418-0250
Methyl dimethoxyacetate 98%
Methyl dimethoxyacetate


Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Boiling Pt: 160…163 °C (760 mmHg)
Density: 1.09 g/cm³
Flash Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008484
CAS Number: 89-91-8
EINECS: 201-950-0
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4070 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty