Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopropylboronic acid pinacol ester 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42859-0010 42859-0050
TS42859-0010EA 87 USD InStock
TS42859-0010 TS42859-0050
Cyclopropylboronic acid pinacol ester 95%
Cyclopropylboronic acid pinacol ester


Formula: C₉H₁₇BO₂
Boiling Pt: 146 °C (760 mmHg)
Density: 0.922 g/cm³
Flash Pt: 40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663847
CAS Number: 126689-01-8
UN: 3272
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty