Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-(+)-Tartaric acid dipotassium salt 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

42799-1000 42799-5000
TS42799-1000EA 56 USD InStock
TS42799-1000 TS42799-5000
L-(+)-Tartaric acid dipotassium salt 97%
L-(+)-Tartaric acid dipotassium salt


Formula: C₄H₄K₂O₆
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013065
CAS Number: 921-53-9
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤4 % (150°C, 4 hr)
Specific optical rotation +25° to +27° (20°C, 589 nm) (c=1,H2O)
Titration with HClO4 ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty