Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Ethoxycarbonyl)phenylboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36779-0010 36779-0050 36779-0250
TS36779-0010EA 59 USD InStock
TS36779-0010 TS36779-0050 TS36779-0250
4-(Ethoxycarbonyl)phenylboronic acid 97%
4-(Ethoxycarbonyl)phenylboronic acid


Formula: C₉H₁₁BO₄
MW: 193.99 g/mol
Melting Pt: 184…189 °C
Storage Temperature: Ambient
MDL Number: MFCD02179441
CAS Number: 4334-88-7
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty