Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Pyridinesulphonic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

13192-1000 13192-5000
TS13192-1000EA 120 USD InStock
TS13192-1000 TS13192-5000
3-Pyridinesulphonic acid 98%
3-Pyridinesulphonic acid


Formula: C₅H₅NO₃S
Melting Pt: >300 °C
Flash Pt: >66 °C
Storage Temperature: Ambient
MDL Number: MFCD00006381
CAS Number: 636-73-7
EINECS: 211-265-9
UN: 2585
ADR: 8,III
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless
Titration with NaOH ≥97.5 %
Elemental Analysis 9.92 to 10.43 % N
Specific optical rotation +5° to +8° (20°C, 589 nm) (c=2, methanol)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty