Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazole-4-boronic acid pinacol ester 97+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

38294-0010 38294-0050 38294-0250
TS38294-0010EA 73 USD InStock
TS38294-0010 TS38294-0050 TS38294-0250
1H-Pyrazole-4-boronic acid pinacol ester 97+%
1H-Pyrazole-4-boronic acid pinacol ester


Formula: C₉H₁₅BN₂O₂
Melting Pt: 125…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03453063
CAS Number: 269410-08-4
Order Now
Specification Test Results
Appearance Off-white to tan Powder
Infrared spectrum Conforms
GC ≥97.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty