Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Iodosobenzoic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44603-0010 44603-0050
TS44603-0010EA 69 USD InStock
TS44603-0010 TS44603-0050
2-Iodosobenzoic acid 98%
2-Iodosobenzoic acid


Formula: C₇H₅IO₃
Melting Pt: 200…204 °C
Storage Temperature: Freezer
MDL Number: MFCD00002401
CAS Number: 304-91-6
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Iodometric 97.5 to 102.5 %
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty